Anti-TFPI2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1248
BSBTPA1248EA 552 EUR
BSBTPA1248
Anti-TFPI2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Tissue factor pathway inhibitor 2 (TFPI2) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: TFPI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR