Anti-S100 beta Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1303
BSBTPA1303EA 552 EUR
BSBTPA1303
Anti-S100 beta Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Protein S100-B(S100B) detection. Tested with WB in Human;Mouse;Rat.

S100 calcium binding protein B or S100B is a protein of the S-100 protein family. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 21q22.3. S100B is a glial-derived protein that is a well-established biomarker for severity of neurological injury and prognosis for recovery. S100 beta is a calcium-binding protein that is expressed at high levels in brain primarily by astrocytes. Addition of the disulfide-bonded dimeric form of S100 beta to primary neuronal and glial cultures and established cell lines induces axonal extension and alterations in astrocyte proliferation and phenotype, but evidence that S100 beta exerts the same effects in vivo has not been presented. Reeves et al.(1994) demonstrated that the same effects of the S100B protein are exerted in vivo. They found that both astrocytosis and neurite proliferation occurred in transgenic mice expressing elevated levels of S100b. They suggested that these transgenic mice represent a useful model for studies of the role of S100B in glial-neuronal interactions in normal development and function of the brain and for analyzing the significance of elevated levels of the protein in Down syndrome and Alzheimer disease.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: S100 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR