Anti-CDC42 Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1366
BSBTPA1366EA 552 EUR
BSBTPA1366
Anti-CDC42 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cell division control protein 42 homolog(CDC42) detection. Tested with WB, IHC-P in Human; Bovin;Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CDC42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovin, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR