Anti-Connexin 32/GJB1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1367
BSBTPA1367EA 552 EUR
BSBTPA1367
Anti-Connexin 32/GJB1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Gap junction beta-1 protein(GJB1) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Connexin 32/GJB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR