Anti-CX3CL1 Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1400
BSBTPA1400EA 552 EUR
BSBTPA1400
Anti-CX3CL1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Fractalkine(CX3CL1) detection. Tested with WB in Human.

  • Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CX3CL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR