Anti-Resistin Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1473
BSBTPA1473EA 552 EUR
BSBTPA1473
Anti-Resistin Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Resistin(RETN) detection. Tested with WB, IHC-P in Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Resistin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR