Anti-AIMP2/p38 Rabbit Polyclonal Antibody
Lieferant: BosterBio
PA1481
BSBTPA1481EA 552 EUR
BSBTPA1481
Anti-AIMP2/p38 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Aminoacyl tRNA synthase complex-interacting multifunctional protein 2(AIMP2) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: AIMP2/p38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat