Anti-Interferon gamma Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1493
BSBTPA1493EA 552 EUR
BSBTPA1493
Anti-Interferon gamma Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Interferon gamma(IFNG) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Interferon gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR