Anti-FOXP3 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1577
BSBTPA1577EA 552 EUR
BSBTPA1577
Anti-FOXP3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Forkhead box protein P3(FOXP3) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR