Anti-Laminin gamma 1 Rabbit Polyclonal Antibody
Lieferant: BosterBio
PA1581
BSBTPA1581EA 552 EUR
BSBTPA1581
Anti-Laminin gamma 1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Laminin subunit gamma-1(LAMC1) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Laminin gamma 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat