Anti-Prostaglandin E Receptor EP1 Rabbit Polyclonal Antibody
Lieferant: BosterBio
PA1583
BSBTPA1583EA 552 EUR
BSBTPA1583
Anti-Prostaglandin E Receptor EP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Prostaglandin E2 receptor EP1 subtype(PTGER1) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Prostaglandin E Receptor EP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human