Anti-GRB7 Polyclonal Antibody

Lieferant: Boster Bio
PA1589-1
BSBTPA1589-1EA 552 EUR
BSBTPA1589-1
Anti-GRB7 Polyclonal Antibody
Antikörper
Polyclonal antibody for GRB7 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. GRB7 information: Molecular Weight: 59681 MW; Subcellular Localization: Cytoplasm. Cell junction, focal adhesion. Cell membrane; Peripheral membrane protein; Cytoplasmic side. Cytoplasmic granule . Cell projection. Predominantly cytoplasmic. Detected in stress granules, where mRNA is stored under stress conditions (By similarity).

Type: Primary
Antigen: GRB7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR