Anti-Kallikrein 1 Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1633
BSBTPA1633EA 552 EUR
BSBTPA1633
Anti-Kallikrein 1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Kallikrein-1(KLK1) detection. Tested with WB, IHC-P in Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Kallikrein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR