Anti-Lamin B2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1636
BSBTPA1636EA 552 EUR
BSBTPA1636
Anti-Lamin B2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Lamin-B2(LMNB2) detection. Tested with WB, IHC-P, IHC-F in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Lamin B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR