Anti-5 Lipoxygenase Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA1644
BSBTPA1644EA 552 EUR
BSBTPA1644
Anti-5 Lipoxygenase Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Arachidonate 5-lipoxygenase(ALOX5) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: 5 Lipoxygenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human