Anti-Monoamine Oxidase B Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1649
BSBTPA1649EA 552 EUR
BSBTPA1649
Anti-Monoamine Oxidase B Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Amine oxidase[flavin-containing] B(MAOB) detection. Tested with WB, IHC-P in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Monoamine Oxidase B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR