Anti-LCN1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1659
BSBTPA1659EA 552 EUR
BSBTPA1659
Anti-LCN1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Lipocalin-1(LCN1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: LCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR