Anti-Kallikrein 1 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA1709
BSBTPA1709EA 552 EUR
BSBTPA1709
Anti-Kallikrein 1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Kallikrein-1 (KLK1) detection. Tested with WB, IHC-P in Mouse.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Kallikrein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse