Anti-Growth hormone Receptor Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA1726
BSBTPA1726EA 552 EUR
BSBTPA1726
Anti-Growth hormone Receptor Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Growth hormone receptor(GHR) detection. Tested with WB in Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Growth hormone Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat