Anti-Caspase-8 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA1735
BSBTPA1735EA 552 EUR
BSBTPA1735
Anti-Caspase-8 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Caspase-8 (CASP8) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Caspase-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human