Anti-FMO3 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1764
BSBTPA1764EA 552 EUR
BSBTPA1764
Anti-FMO3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Dimethylaniline monooxygenase[N-oxide-forming] 3 (FMO3) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: FMO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR