Anti-PERK Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA2026
BSBTPA2026EA 552 EUR
BSBTPA2026
Anti-PERK Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Eukaryotic translation initiation factor 2-alpha kinase 3 (EIF2AK3) detection. Tested with WB, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: PERK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR