Anti-Cyclophilin B Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA2151
BSBTPA2151EA 552 EUR
BSBTPA2151
Anti-Cyclophilin B Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Peptidyl-prolyl cis-trans isomerase B(PPIB) detection. Tested with WB, IHC-P in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Cyclophilin B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat