Anti-Fibulin 2 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA2159
BSBTPA2159EA 552 EUR
BSBTPA2159
Anti-Fibulin 2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Fibulin-2(FBLN2) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Fibulin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat