Anti-LAT Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA2161
BSBTPA2161EA 552 EUR
BSBTPA2161
Anti-LAT Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Linker for activation of T-cells family member 1(LAT) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR