Anti-GLUT8 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PA2165-10UG
BSBTPA2165-10UGEA 174 EUR
BSBTPA2165-10UG
Anti-GLUT8 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Solute carrier family 2, facilitated glucose transporter member 8 (SLC2A8) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 μg/ml.
Type: Primary
Antigen: GLUT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat