Anti-alpha 2a Adrenergic Receptor Rabbit Antibody

Lieferant: Boster Bio
PA2197
BSBTPA2197EA 552 EUR
BSBTPA2197
Anti-alpha 2a Adrenergic Receptor Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Alpha-2A adrenergic receptor(ADRA2A) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: ADRA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR