Anti-Cdc6 Rabbit Antibody

Lieferant: Boster Bio
PA2199
BSBTPA2199EA 552 EUR
BSBTPA2199
Anti-Cdc6 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cell division control protein 6 homolog(CDC6) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CDC6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR