Anti-Connexin 45/GJA7 Rabbit Antibody

Lieferant: Boster Bio
PA2206
BSBTPA2206EA 552 EUR
BSBTPA2206
Anti-Connexin 45/GJA7 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Gap junction gamma-1 protein(GJC1) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Connexin 45/GJA7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR