Anti-Sumo 1 Rabbit Antibody

Lieferant: Boster Bio
PA2220
BSBTPA2220EA 552 EUR
BSBTPA2220
Anti-Sumo 1 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Small ubiquitin-related modifier 1(SUMO1) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: SUMO 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR