Anti-CD47 Rabbit Antibody

Lieferant: Boster Bio
PA2223
BSBTPA2223EA 0 EUR
BSBTPA2223
Anti-CD47 Rabbit Antibody
Antikörper
Type: Primary
Antigen: CD47
Clonality:
Clone:
Conjugation:
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR