Anti-IRF1 Rabbit Antibody

Lieferant: Boster Bio
PA2237
BSBTPA2237EA 552 EUR
BSBTPA2237
Anti-IRF1 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Interferon regulatory factor 1(IRF1) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: IRF1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR