Anti-RIP3 Rabbit Antibody

Lieferant: Boster Bio
PA2242
BSBTPA2242EA 552 EUR
BSBTPA2242
Anti-RIP3 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Receptor-interacting serine/threonine-protein kinase 3(RIPK3) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: RIPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR