Anti-NCSTN Rabbit Antibody

Lieferant: Boster Bio
PA2250
BSBTPA2250EA 552 EUR
BSBTPA2250
Anti-NCSTN Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Nicastrin(NCSTN) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: NCSTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR