Anti-PRMT4 Rabbit Antibody

Lieferant: Boster Bio
PA2263
BSBTPA2263EA 552 EUR
BSBTPA2263
Anti-PRMT4 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Histone-arginine methyltransferase CARM1(CARM1) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CARM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR