Anti-Huntingtin Rabbit Antibody

Lieferant: Boster Bio
PA2268
BSBTPA2268EA 552 EUR
BSBTPA2268
Anti-Huntingtin Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Huntingtin(HTT) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR