Anti-CSEN Rabbit Antibody

Lieferant: Boster Bio
PA2272
BSBTPA2272EA 552 EUR
BSBTPA2272
Anti-CSEN Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Calsenilin(KCNIP3) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CSEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR