Anti-Rab3C Rabbit Antibody

Lieferant: Boster Bio
PA2279
BSBTPA2279EA 552 EUR
BSBTPA2279
Anti-Rab3C Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Ras-related protein Rab-3C(RAB3C) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: RAB3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR