Anti-ST5 Rabbit Antibody

Lieferant: Boster Bio
PA2287
BSBTPA2287EA 552 EUR
BSBTPA2287
Anti-ST5 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Suppression of tumorigenicity 5 protein(ST5) detection. Tested with WB in Human;Mouse;Rat.

Suppression of tumorigenicity 5 is a protein that in humans is encoded by the ST5 gene. This gene is mapped to 11p15.4. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: ST5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR