Anti-Mucin 5AC Rabbit Antibody

Lieferant: Boster Bio
PA2303
BSBTPA2303EA 552 EUR
BSBTPA2303
Anti-Mucin 5AC Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Mucin-5AC(MUC5AC) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: MUC5AC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR