Anti-TNFSF11 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB10015
BSBTPB10015EA 552 EUR
BSBTPB10015
Anti-TNFSF11 Rabbit Polyclonal Antibody
Antikörper
Receptor activator of nuclear factor kappa-B ligand (RANKL), also known as tumor necrosis factor ligand superfamily member 11 (TNFSF11), is a protein that in humans is encoded by the TNFSF11 gene. This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. It is mapped to chromosome13q14.11. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregnancy. This gene may play an important role in enhanced bone-resorption in humoral hypercalcemia of malignancy.
1. Asselin-Labat, M.-L., Vaillant, F., Sheridan, J. M., Pal, B., Wu, D., Simpson, E. R. Yasuda, H.; Smyth, G. K.; Martin, T. J.; Lindeman, G. J.; Visvader, J. E.: Control of mammary stem cell function by steroid hormone signalling. Nature 465: 798-802, 2010.
2. Luan, X., Lu, Q., Jiang, Y., Zhang, S., Wang, Q., Yuan, H., Zhao, W., Wang, J., Wang, X.Crystal structure of human RANKL complexed with its decoy receptor osteoprotegerin. J Immun. 189: 245-252, 2012.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.