Anti-AKR1C1/C2 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB10036
BSBTPB10036EA 552 EUR
BSBTPB10036
Anti-AKR1C1/C2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Aldo-keto reductase family 1 member C1/C2(AKR1C1/C2) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: AKR1C1/C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR