Anti-CYP3A4 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB10055
BSBTPB10055EA 552 EUR
BSBTPB10055
Anti-CYP3A4 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cytochrome P450 3A4(CYP3A4) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: CYP3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR