Anti-GNB1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB10067
BSBTPB10067EA 552 EUR
BSBTPB10067
Anti-GNB1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1(GNB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: GNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR