Anti-NR2F6 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB10081
BSBTPB10081EA 552 EUR
BSBTPB10081
Anti-NR2F6 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Nuclear receptor subfamily 2 group F member 6(NR2F6) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: NR2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR