Anti-PSMA3 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB10089
BSBTPB10089EA 552 EUR
BSBTPB10089
Anti-PSMA3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Proteasome subunit alpha type-3(PSMA3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat