Anti-COX1/Cyclooxygenase 1 Polyclonal Antibody

Lieferant: Boster Bio
PB9002
BSBTPB9002EA 552 EUR
BSBTPB9002
Anti-COX1/Cyclooxygenase 1 Polyclonal Antibody
Antikörper
Polyclonal antibody for COX 1/PTGS1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. COX 1/PTGS1 information: Molecular Weight: 68686 MW; Subcellular Localization: Microsome membrane; Peripheral membrane protein. Endoplasmic reticulum membrane; Peripheral membrane protein.

Type: Primary
Antigen: COX1/Cyclooxygenase 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR