Anti-AKT2 Polyclonal Antibody

Lieferant: Boster Bio
PB9043
BSBTPB9043EA 552 EUR
BSBTPB9043
Anti-AKT2 Polyclonal Antibody
Antikörper
Polyclonal antibody for AKT2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. AKT2 information: Molecular Weight: 55769 MW; Subcellular Localization: Cytoplasm. Nucleus. Cell membrane; Peripheral membrane protein. Localizes within both nucleus and cytoplasm of proliferative primary myoblasts and mostly within the nucleus of differentiated primary myoblasts. By virtue of the N-terminal PH domain, is recruited to sites of the plasma membrane containing increased PI(3,4,5)P3 or PI(3,4)P2, cell membrane targeting is also facilitared by interaction with CLIP3; Tissue Specificity: Expressed in all cell types so far analyzed.

Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR