Anti-NF-kB p65 Polyclonal Antibody

Lieferant: Boster Bio
PB9087
BSBTPB9087EA 552 EUR
BSBTPB9087
Anti-NF-kB p65 Polyclonal Antibody
Antikörper
Polyclonal antibody for NFkB p65/RELA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. NFkB p65/RELA information: Molecular Weight: 60219 MW; Subcellular Localization: Nucleus. Cytoplasm. Colocalized with DDX1 in the nucleus upon TNF-alpha induction (By similarity). Nuclear, but also found in the cytoplasm in an inactive form complexed to an inhibitor (I-kappa-B). Colocalizes with GFI1 in the nucleus after LPS stimulation.

Type: Primary
Antigen: NF-kB p65
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR