Anti-ATF1 Polyclonal Antibody

Lieferant: BosterBio
PB9130
BSBTPB9130EA 552 EUR
BSBTPB9130
Anti-ATF1 Polyclonal Antibody
Antikörper
Polyclonal antibody for 19261910 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. 19261910 information: Molecular Weight: 29233 MW; Subcellular Localization: Nucleus.

Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR